missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF750-SP
This item is not returnable.
View return policy
Description
CD6 Polyclonal specifically detects CD6 in Mouse samples. It is validated for Western Blot, Adhesion Blockade.
Specifications
| CD6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 923 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Adhesion Blockade | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Adhesion Blockade | |
| Q91WN5 | |
| CD6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD6 Leu18-Gly396 Accession # Q91WN5 | |
| 25 μg | |
| Primary | |
| Detects mouse CD6 in direct ELISAs and Western blots. In Western blots, approximately 1% cross-reactivity with recombinant human CD6 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction