missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD6 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marca: R&D Systems AF750
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD6 Polyclonal specifically detects CD6 in Mouse samples. It is validated for Western Blot, Adhesion Blockade.
Especificaciones
| CD6 | |
| Polyclonal | |
| Unconjugated | |
| Q91WN5 | |
| CD6 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD6 Leu18-Gly396 Accession # Q91WN5 | |
| 100 μg | |
| Primary | |
| Detects mouse CD6 in direct ELISAs and Western blots. In Western blots, approximately 1% cross-reactivity with recombinant human CD6 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Adhesion Blockade | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Adhesion Blockade | |
| CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 923 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido