missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL8/MCP-2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems BAF790
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCL8/MCP-2 Polyclonal specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Western Blot.
Especificaciones
| CCL8/MCP-2 | |
| Polyclonal | |
| Biotin | |
| Q9Z121 | |
| CCL8 | |
| E. coli-derived recombinant mouse CCL8/MCP-2 Glu20-Pro97 Accession # Q9Z121 | |
| 50 μg | |
| Primary | |
| Detects mouse CCL8/MCP-2 in Western blots. In Western blots, less than 1% cross-reactivity with Brucella melitensis BcsP31 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6355 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido