missing translation for 'onlineSavingsMsg'
Learn More

Mouse Caspase-11/Caspase-4 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18726488
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18726488 25 μg 25 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18726488 Proveedor R&D Systems N.º de proveedor MAB86481SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

Caspase-11/Caspase-4 Monoclonal specifically detects Caspase-11/Caspase-4 in Mouse samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Caspase-11/Caspase-4
Aplicaciones Immunocytochemistry
Clasificación Monoclonal
Clon 919916
Conjugado Unconjugated
Dilución Immunocytochemistry 8-25 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P70343
Símbolos de los genes Casp4
Especie del huésped Rat
Inmunógeno E. coli-derived recombinant mouse Caspase-11/Caspase-4 Pro81-Glu266 Accession # P70343
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12363
Especificidad de la prueba Detects mouse Caspase-11/Caspase-4 in direct ELISA.
Reconstitución Reconstitute at 0.5 mg/mL in sterile PBS.
Especies diana Mouse
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Formulario Purified
Isotype IgG2b
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.