missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB50201
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BMP-4 Monoclonal specifically detects BMP-4 in Mouse samples. It is validated for Neutralization.
Especificaciones
| BMP-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 652 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 871317 | |
| Neutralization 0.4-2.4 ug/mL | |
| P21275 | |
| Bmp4 | |
| Chinese hamster ovary cell line CHO-derived mature mouse BMP-4 Ser293-Arg408 Accession # P21275 | |
| 100 μg | |
| Primary | |
| Detects mouse BMP-4 in direct ELISAs. | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido