missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-1/CD80 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Marca: R&D Systems AF740-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
B7-1/CD80 Polyclonal specifically detects B7-1/CD80 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Especificaciones
| B7-1/CD80 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 941 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF, ELISA | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.15-0.6 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Q00609 | |
| CD80 | |
| Mouse myeloma cell line NS0-derived recombinant mouse B7-1/CD80 Asp37-Lys245 Accession # Q00609 | |
| 25 μg | |
| Primary | |
| Detects mouse B7-1/CD80 in ELISAs and Western blots. | |
| Mouse | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido