Learn More
ZAK, Mouse, Clone: 4C4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ZAK.
Marca: Abnova H00051776-M07A.200uL
Descripción
This gene is a member of the MAPKKK family of signal transduction molecules and encodes a protein with an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif (SAM). This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: MSSLGASFVQIKFDDLQFFENCGGGSFGSVYRAKWISQDKEVAVKKLLKIEKEAEILSVLSHRNIIQFYGVILEPPNYGIVTEYASLGSLYDYINSNRSEEMDMDHIMTWATDVAKGMHYEspecificaciones
| ZAK | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC001401 | |
| ZAK | |
| ZAK (AAH01401,1 a.a. ∽ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51776 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4C4 | |
| Mouse monoclonal antibody raised against a partial recombinant ZAK. | |
| ZAK | |
| AZK/MLK7/MLT/MLTK/MRK/mlklak | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.