Learn More
TNFRSF14, Mouse, Clone: ANC3B7, Abnova™
Descripción
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to this receptor protein has been shown to be part of the viral entry mechanism. The cytoplasmic region of this receptor was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | TNFRSF14 |
| Aplicaciones | ELISA, Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | ANC3B7 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against recombinant TNFRSF14. |
| Dilución | The optimal working dilution should be determined by the end user. |
| Formulación | In 50mM sodium phosphate buffer, 100mM potassium chloride, 150mM NaCl, pH 7.5 (0.5mg/mL gentamicin sulfate) |
| génica | TNFRSF14 |
| Alias de gen | ATAR/HVEA/HVEM/LIGHTR/TR2 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.