Learn More
SUZ12, Mouse, Clone: 3D10, Abnova™
Mouse monoclonal antibody raised against recombinant SUZ12.
Marca: Abnova MAB10328.100uL
Descripción
This zinc finger gene has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region. [provided by RefSeq
Especificaciones
| SUZ12 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| SUZ12 | |
| SUZ12 | |
| Recombinant protein corresponding to human SUZ12. | |
| RUO | |
| 23512 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 3D10 | |
| Mouse monoclonal antibody raised against recombinant SUZ12. | |
| In ascites (0.03% sodium azide) | |
| CHET9/JJAZ1/KIAA0160 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.