missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAH2, Mouse, Clone: 24E6H3, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of SIAH2.
Marca: Abnova MAB2298.500uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| SIAH2 | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (1:5-1:10) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:40-1:50) Immunohistochemistry (Frozen sections) (1:40-1:50) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 500 μL | |
| Primary | |
| Drosophila, Human, Pig | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 24E6H3 | |
| Mouse monoclonal antibody raised against synthetic peptide of SIAH2. | |
| In tissue culture supernatant (0.09% sodium azide) | |
| A synthetic peptide corresponding to amino acids 1-100 of drosophila SIAH2. | |
| RUO | |
| This antibody is specific to human SIAH2 and Drosophila SINA in proliferating cells and stem cells. This antibody does not detect Drosophila SINAH or normal, nondividing cells. | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido