Learn More
PTPN6, Mouse, Clone: M160, Abnova™
Descripción
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. N-terminal part of this PTP contains two tandem Src homolog (SH2) domains, which act as protein phospho-tyrosine binding domains, and mediate the interaction of this PTP with its substrates. This PTP is expressed primarily in hematopoietic cells, and functions as an important regulator of multiple signaling pathways in hematopoietic cells. This PTP has been shown to interact with, and dephosphorylate a wide spectrum of phospho-proteins involved in hematopoietic cell signaling. Multiple alternatively spliced variants of this gene, which encode distinct isoforms, have been reported. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | PTPN6 |
| Aplicaciones | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | M160 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against partial recombinant PTPN6. |
| Dilución | ELISA (1:2000) Immunocytochemistry (1:100) Immunoprecipitation (5-10 ul) Western Blot (1:500) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) |
| génica | PTPN6 |
| Alias de gen | HCP/HCPH/HPTP1C/PTP-1C/SH-PTP1/SHP-1/SHP-1L/SHP1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.