Learn More
PRDX5, Mouse, Clone: 9g056, Abnova™
Mouse monoclonal antibody raised against full-length recombinant PRDX5.
Marca: Abnova MAB8843.100uL
Descripción
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in different tissues under normal conditions and during inflammatory processes. This protein interacts with peroxisome receptor 1. The crystal structure of this protein in its reduced form has been resolved to 1.5 angstrom resolution. This gene uses alternate in-frame translation initiation sites to generate mitochondrial or peroxisomal/cytoplasmic forms. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Especificaciones
| PRDX5 | |
| Monoclonal | |
| 1 ug/uL | |
| Mouse monoclonal antibody raised against full length recombinant PRDX5. | |
| In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
| ACR1/AOEB166/B166/MGC117264/MGC142283/MGC142285/PLP/PMP20/PRDX6/PRXV/SBBI10 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 25824 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 9g056 | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| PRDX5 | |
| PRDX5 | |
| Recombinant protein corresponding to full length of human PRDX5. | |
| 100 μL | |
| Primary | |
| This antibody recognizes recombinant protein PRDX5. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.