missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-B Mouse anti-Human, Clone: 108132, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Marca: R&D Systems MAB220-500
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot.Especificaciones
| PDGF-B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B chain, Becaplermin, C-Sis, FLJ12858, IBGC5, PDGF subunit B, PDGF2, PDGF-2, PDGFB, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SIS, SSV | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5155 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 108132 | |
| Western Blot 1 ug/mL | |
| P01127 | |
| PDGFB | |
| E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127 | |
| 500 μg | |
| Primary | |
| Detects human PDGF-B subunit in direct ELISAs and Western blots. In direct ELISAs, shows approximately 40% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhCTGF, rhLDGF, rhPDGF-AA, recombinant rat (rr) PDGF-AA, rrPDGF-AB, rrPDGF-BB, rhPDGF-C, rhPDGF-D, rhPlGF, rhVEGF, rhVEGF-C, or rhVEGF-D. | |
| Human | |
| Supernatant |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?