Learn More
NUDC, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant NUDC.
Marca: Abnova H00010726-A01.50uL
Descripción
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein (see DYNC1H1; MIM 600112) (Aumais et al., 2003 [PubMed 12679384]).[supplied by OMIM
Sequence: MGGEQEEERFDGMLLAMAQQHEGGVQELVNTFFSFLRRKTDFFIGGEEGMAEKLITQTFSHHNQLAQKTRREKRARQEAERREKAERAARLAKEAKSETSGPQIKELTDEEAERLQLEIDQKKDAENHEAQLKNGSLDSPGKQDTEEDEGEDEKDKGKLKPNLGNGADLPNYRWTQTLSELDLAVPFCVNFRLKGKDMVVDIQRRHLRVGLKGQPAIIDGELYNEVKVEESSWLIEDGKVVTVHLEKINKMEWWSRLVSSDPEINTKKINPENSKLSDLDSETRSMVEKMMYDQRQKSMGLPTSDEQKKQEILKKFMDQHPEMDFSKAKFNEspecificaciones
| NUDC | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant NUDC. | |
| NUDC | |
| HNUDC/MNUDC/NPD011 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA | |
| Unconjugated | |
| 50% glycerol | |
| BC002399 | |
| NUDC | |
| NUDC (AAH02399, 1 a.a. ∽ 331 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 10726 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only