Learn More
JMJD2A, Mouse, Clone: 5H1, Abnova™
Mouse monoclonal antibody raised against partial recombinant JMJD2A.
Marca: Abnova MAB10708.100uL
Descripción
This gene is a member of the Jumonji domain 2 (JMJD2) family and encodes a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This nuclear protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor. [provided by RefSeq
Especificaciones
| JMJD2A | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| JMJD2A | |
| JMJD2A | |
| Recombinant protein corresponding to human JMJD2A. | |
| RUO | |
| 9682 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 5H1 | |
| Mouse monoclonal antibody raised against partial recombinant JMJD2A. | |
| In ascites (0.03% sodium azide) | |
| JHDM3A/JMJD2/KDM4A/KIAA0677 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.