Learn More
RON (CD136) Mouse anti-Human, Unlabeled, Clone: Zt/g4, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 565691
Descripción
The Zt/g4 monoclonal antibody specifically binds to RON, which is also known as CD136, Macrophage-stimulating protein receptor (MSP-Receptor, MSP-R), Macrophage stimulating 1 receptor (c-met-related tyrosine kinase) (MST1R), or Protein-tyrosine kinase 8 (PTK8). RON is a type I transmembrane glycoprotein that belongs to the receptor tyrosine kinase family. It is expressed on cells as a disulfide-linked heterodimer. RON is expressed on macrophages, keratinocytes, erythroblasts and epithelial cells from the skin, lung, kidney, liver, intestine, colon, and bone marrow. RON reportedly regulates many physiological processes including the survival, migration, differentiation, and effector functions of a variety of cell types including epithelial cells, and macrophages. RON binds Macrophage stimulating protein (MSP). Overexpression of RON may contribute to the malignant transformation of epithelial cells.
Especificaciones
| RON (CD136) | |
| Monoclonal | |
| 0.5mg/mL | |
| MSP Receptor; MSP-R; MST1R; p185-Ron; PTK8 protein tyrosine kinase 8; PTK8 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry | |
| Zt/g4 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human RON Transfected Cell Line | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.