Learn More
INHBA, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant INHBA.
Marca: Abnova H00003624-A01.50uL
Descripción
The inhibin beta A subunit joins the alpha subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. Because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. Furthermore, the beta A subunit forms a homodimer, activin A, and also joins with a beta B subunit to form a heterodimer, activin AB, both of which stimulate FSH secretion. Finally, it has been shown that the beta A subunit mRNA is identical to the erythroid differentiation factor subunit mRNA and that only one gene for this mRNA exists in the human genome. [provided by RefSeq
Sequence: MPLLWLRGFLLASCWIIVRSSPTPGSEGHSAAPDCPSCALAALPKDVPNSQPEMVEAVKKHILNMLHLKKRPDVTQPVPKAALLNAIRKLHVGKVGENGYVEIEDDIGRRAEMNELMEQTSEIITFAESGTARKTLHFEISKEGSDLSVVERAEVWLFLKVPKANRTRTKVTIRLFQQQKHPQGSLDTGEEAEEVGLKGERSELLLSEKVVDARKSTWHVFPVSSSIQRLLDQGKSSLDVRIACEQCQESGASLVLLGKKKKKEEEGEGKKKGGGEGGAGADEEKEQSHRPFLMLQARQSEDHPHRRRRRGLECDGKVNICCKKQFFVSFKDIGWNDWIIAPSGYHANYCEGECPSHIAGTSGSSLSFHSTVINHYRMRGHSPFANLKSCCVPTKLRPMSMLYYDDGQNIIKKDIQNMIVEECGCSEspecificaciones
| INHBA | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant INHBA. | |
| INHBA | |
| EDF/FRP | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC007858 | |
| INHBA | |
| INHBA (AAH07858.1, 1 a.a. ∽ 426 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 3624 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only