Learn More
Invitrogen™ Mouse anti-Human Kappa Light Chain Secondary Antibody
Código de producto. 11545020
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
11545020 100 μg 100 microgramos
1 options
Código de producto. 11545020
Marca: Invitrogen™ SA119155
Mouse Monoclonal Secondary Antibody
This antibody reacts with kappa light chains (22.5 kDa). Material immunoprecipitated from human serum with the antibody MEM-09 consists of IgG and traces of IgM.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Human Kappa Light Chain
Aplicaciones ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Clasificación Monoclonal
Clon MEM-09
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 15mM sodium azide; pH 7.4
Especie del huésped Mouse
Inmunógeno Crude thymus membrane fraction.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Human
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG1