Learn More
CD16 Mouse anti-Human, Brilliant Violet 711, Clone: B73.1, BD Optibuild™
Descripción
The B73.1 monoclonal antibody specifically binds to CD16, the 50-70 kDa low affinity receptor for IgG, IgG Fc receptor III (Fc γRIII) . CD16 is expressed on NK cells, neutrophils, and on a subset of T cells from certain individuals. The B73.1 antibody binds to CD16-positive neutrophils with lower intensity when compared with some other CD16-specific antibodies. A variable number of CD16-positive lymphocytes coexpress either the CD57 antigen or low-density CD8 antigen or both. The B73.1 antibody can block Fc receptor functions mediated by CD16.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Especificaciones
Especificaciones
| Antígeno | CD16 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | B73.1 |
| Concentración | 0.2mg/mL |
| Conjugado | Brilliant Violet 711 |
| Descripción | IgG Fc receptor III; IGFR3; FCG3; FCGR3; FCGRIII; FcγRIII |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human NK Cells |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.