Learn More
HGH Mouse anti-Human, Clone: 4C1C12, Abnova™
Mouse monoclonal antibody raised against full-length native HGH.
Marca: Abnova MAB1578.1mg
Descripción
The protein encoded by this gene is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The gene, along with four other related genes, is located at the growth hormone locus on chromosome 17 where they are interspersed in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. The five genes share a remarkably high degree of sequence identity. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. This particular family member is expressed in the pituitary but not in placental tissue as is the case for the other four genes in the growth hormone locus. Mutations in or deletions of the gene lead to growth hormone deficiency and short stature. [provided by RefSeq
Especificaciones
| HGH | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| GH1 | |
| GH1 | |
| Native purified human growth hormone. | |
| RUO | |
| 2688 | |
| Liquid |
| ELISA | |
| 4C1C12 | |
| Mouse monoclonal antibody raised against full length native HGH. | |
| In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide) | |
| GH/GH-N/GHN/hGH-N | |
| Mouse | |
| 1 mg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.