Learn More
GYPA, Mouse, Clone: CMRF14, Abnova™
Descripción
Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. In addition to the M or N and S or s antigens that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta, as well as Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | GYPA |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | CMRF14 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against full length native GYPA. |
| Dilución | Flow cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS |
| génica | GYPA |
| Alias de gen | CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.