Learn More
Mouse anti-Goat IgG whole molecule, Polyclonal, Secondary Antibody, Abnova™
Código de producto. 16234630
Change view
Click to view available options
Cantidad:
2 mL
Tamaño de la unidad:
2 ml
16234630 2 mL 2 ml
1 options
Código de producto. 16234630
Marca: Abnova PAB9210.2mL
Especificaciones
Antígeno Mouse anti-Goat IgG whole molecule
Aplicaciones ELISA, Immunohistochemistry, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Mouse anti-Goat IgG recognizes Goat IgG whole molecule. This secondary antibody was purified using delipidation and defibrination.
Dilución The optimal working dilution should be determined by the end user.
Formulación Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2
Especie del huésped Mouse
Inmunógeno Goat IgG whole molecule
Método de purificación This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against goat IgG and goat serum.
Cantidad 2 mL
Primario o secundario Secondary
Especies diana Goat
Contenido y almacenamiento Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Formulario Lyophilized
Mostrar más Mostrar menos
For Research Use Only