Learn More
F5 Mouse anti-Human, Monoclonal Antibody, Abnova™
Descripción
This gene encodes an essential cofactor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The activated protein is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in this gene result in either an autosomal recessive hemorrhagic diathesis or an autosomal dominant form of thrombophilia, which is known as activated protein C resistance. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | F5 |
| Aplicaciones | Inhibition Assays, Radioimmune Assays (RIA) |
| Clasificación | Monoclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against native F5. |
| Dilución | Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS (50% glycerol) |
| génica | F5 |
| Alias de gen | FVL/PCCF |
| Símbolos de los genes | F5 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.