Learn More
ELANE Mouse anti-Human, Monoclonal Antibody, Abnova™
Descripción
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins. The product of this gene hydrolyzes proteins within specialized neutrophil lysosomes, called azurophil granules, as well as proteins of the extracellular matrix following the protein's release from activated neutrophils. The enzyme may play a role in degenerative and inflammatory diseases by its proteolysis of collagen-IV and elastin of the extracellular matrix. This protein degrades the outer membrane protein A (OmpA) of E. coli as well as the virulence factors of such bacteria as Shigella, Salmonella and Yersinia. Mutations in this gene are associated with cyclic neutropenia and severe congenital neutropenia (SCN). This gene is clustered with other serine protease gene family members, azurocidin 1 and proteinase 3 genes, at chromosome 19pter. All 3 genes are expressed coordinately and their protein products are packaged together into azurophil granules during neutrophil differentiation. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | ELANE |
| Aplicaciones | ELISA, Radioimmune Assays (RIA) |
| Clasificación | Monoclonal |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against native ELANE. |
| Dilución | The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.4 |
| génica | ELA2 |
| Alias de gen | GE/HLE/HNE/NE/PMN-E |
| Símbolos de los genes | ELA2 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.