Learn More
DPP4, Mouse, Clone: BA5b, Abnova™
Mouse monoclonal antibody raised against DPP4.
Marca: Abnova MAB8513.100ug
Descripción
The protein encoded by this gene is identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. It is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. [provided by RefSeq
Especificaciones
| DPP4 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| DPP4 | |
| DPP4 | |
| A human T cell clone. | |
| RUO | |
| 1803 | |
| Human | |
| Liquid |
| Flow Cytometry | |
| BA5b | |
| Mouse monoclonal antibody raised against DPP4. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| ADABP/ADCP2/CD26/DPPIV/TP103 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody is a peptidase expressed on mature thymocytes, T cells (especially activated), B cells, NK cells and macrophages. | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.