Learn More
CTSD, Mouse, Clone: 4G2, Abnova™
Mouse monoclonal antibody raised against partial recombinant CTSD.
Marca: Abnova MAB1083.100uL
Descripción
This gene encodes a lysosomal aspartyl protease composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. This proteinase, which is a member of the peptidase C1 family, has a specificity similar to but narrower than that of pepsin A. Transcription of this gene is initiated from several sites, including one which is a start site for an estrogen-regulated transcript. Mutations in this gene are involved in the pathogenesis of several diseases, including breast cancer and possibly Alzheimer disease. [provided by RefSeq
Especificaciones
| CTSD | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| CTSD | |
| CLN10/CPSD/MGC2311 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 4G2 | |
| Mouse monoclonal antibody raised against partial recombinant CTSD. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| NM_001909 | |
| CTSD | |
| Recombinant protein corresponding to amino acids 21-412 of human CTSD. | |
| RUO | |
| 1509 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.