Learn More
CGB, Mouse, Clone: HCG-8, Abnova™
Descripción
This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 3 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | CGB |
| Aplicaciones | Immunohistochemistry (PFA fixed) |
| Clasificación | Monoclonal |
| Clon | PC-2 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against native CGB. |
| Dilución | Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulación | Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) |
| génica | CGB |
| Alias de gen | CGB3/hCGB |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.