Learn More
CGA, Mouse, Clone: 1C5, Abnova™
Mouse monoclonal antibody raised against full-length native CGA.
Marca: Abnova MAB1557.1mg
Descripción
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq
Especificaciones
| CGA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CGA | |
| CGA | |
| Native purified human CGA. | |
| RUO | |
| 1081 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA | |
| 1C5 | |
| Mouse monoclonal antibody raised against full length native CGA. | |
| In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide) | |
| CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA | |
| Mouse | |
| 1 mg | |
| Primary | |
| This antibody react with Alpha-hCG, LH, FSH, TSH. No cross reactivity with Beta-hCG. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.