Learn More
CEBPB, Mouse, Clone: 3H9, Abnova™
Descripción
The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related proteins CEBP-alpha, CEBP-delta, and CEBP-gamma. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses and has been shown to bind to the IL-1 response element in the IL-6 gene, as well as to regulatory regions of several acute-phase and cytokine genes. In addition, the encoded protein can bind the promoter and upstream element and stimulate the expression of the collagen type I gene. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | CEBPB |
| Aplicaciones | ELISA, Flow Cytometry, Western Blot |
| Clasificación | Monoclonal |
| Clon | 3H9 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against partial recombinant CEBPB. |
| Dilución | ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
| Formulación | In ascites (0.03% sodium azide) |
| génica | CEBPB |
| Alias de gen | C/EBP-beta/CRP2/IL6DBP/LAP/MGC32080/NF-IL6/TCF5 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.