Learn More
CDH1, Mouse, Clone: 7A2, Abnova™
Descripción
This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | CDH1 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 7A2 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against partial recombinant CDH1. |
| Dilución | ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
| Formulación | In ascites (0.03% sodium azide) |
| génica | CDH1 |
| Alias de gen | Arc-1/CD324/CDHE/ECAD/LCAM/UVO |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.