Learn More
CD4, Mouse, SPRD, Clone: RFT-4g, Abnova™
Mouse monoclonal antibody raised against native CD4.
Marca: Abnova MAB6016.100Reactions
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| CD4 | |
| Monoclonal | |
| SPRD | |
| Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
| CD4 | |
| CD4 | |
| Native purified CD4 from human thymocytes and E-rosetted lymphocytes. | |
| RUO | |
| 920 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| RFT-4g | |
| Mouse monoclonal antibody raised against native CD4. | |
| In PBS (0.09% sodium azide) | |
| CD4mut | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Specificity human CD4 | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |