Learn More
CD39 Mouse anti-Human, PerCP-Cy5.5, Clone: TU66, BD
Descripción
The TU66 monoclonal antibody reacts with human CD39 also known as ectonucleoside triphosphate diphosphohydrolase 1 (ENTPD1) an ectoenzyme that degrades ATP to AMP. It is a member of the family of ectonucleoside triphosphate dihydrolases (E-NTPDases) known to be involved in regulation of extracellular nucleotide catabolism, controlling the extracellular nucleoside triphosphate pool (NTP). CD39 is expressed on a subset of T cells, B cells and dendritic cells with weak staining of monocytes and granulocytes. Recently, CD39 has been found to be expressed primarly by immune-suppressive Foxp3(+) regulatory T (Treg) cells in both human and mice. In humans, CD39 is restricted to a subset of Foxp3+ regulatory effector/memory-like T cells. In mice, the enzyme is present on most if not all CD4+CD25+ cells and CD39 expression is driven by Foxp3. It is thought that CD39 allows Treg cells to enter inflamed areas where high levels of ATP are present.
Especificaciones
Especificaciones
| Antígeno | CD39 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | TU66 |
| Conjugado | PerCP-Cyanine5.5 |
| Descripción | ENTPD1; NTPDase-1; Ecto-ATPase 1; Ecto-ATPDase 1 |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Cantidad | 100 Tests |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.