missing translation for 'onlineSavingsMsg'
Learn More

CD209, Mouse, Clone: 5D7, Abnova™

Código de producto. 16045746
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16045746 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16045746

Marca: Abnova MAB2748.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse monoclonal antibody raised against recombinant CD209.

This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants

Especificaciones

Antígeno CD209
Aplicaciones Immunohistochemistry, Western Blot
Clasificación Monoclonal
Clon 5D7
Conjugado Unconjugated
Descripción Mouse monoclonal antibody raised against recombinant CD209.
Dilución Western Blot (1-2 ug/mL) Immunohistochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In PBS (0.02% sodium azide)
génica CD209
Alias de gen CDSIGN/CLEC4L/DC-SIGN/DC-SIGN1/MGC129965
Símbolos de los genes CD209
Especie del huésped Mouse
Inmunógeno Recombinant His fusion protein corresponding to extracellular region of human CD209.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 30835
Especies diana Human
Contenido y almacenamiento Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.