Learn More
CCL5, Mouse, Clone: 8.PP.11, Abnova™
Mouse monoclonal antibody raised against CCL5.
Marca: Abnova MAB8063.50ug
Descripción
This gene is one of several CC cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. [provided by RefSeq]
Especificaciones
| CCL5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (2-4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CCL5 | |
| CCL5 | |
| Human CCL5. | |
| RUO | |
| 6352 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| ELISA, Immunohistochemistry, Inhibition Assays | |
| 8.PP.11 | |
| Mouse monoclonal antibody raised against CCL5. | |
| In PBS, pH 7.2 (40% glycerol) | |
| D17S136E/MGC17164/RANTES/SCYA5/SISd/TCP228 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.