Learn More
BMX, Mouse, Clone: 1C6, Abnova™
Descripción
This gene encodes a nonreceptor tyrosine kinase belonging to the Tec kinase family. The protein contains a PH-like domain, which mediates membrane targeting by binding to phosphatidylinositol 3,4,5-triphosphate (PIP3), and a SH2 domain that binds to tyrosine-phosphorylated proteins and functions in signal transduction. The protein is implicated in several signal transduction pathways including the Stat pathway, and regulates differentiation and tumorigenicity of several types of cancer cells. Multiple alternatively spliced variants, encoding the same protein, have been identified
Especificaciones
Especificaciones
| Antígeno | BMX |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1C6 |
| Conjugado | Unconjugated |
| Descripción | Mouse monoclonal antibody raised against partial recombinant BMX. |
| Dilución | ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
| Formulación | In ascites (0.03% sodium azide) |
| génica | BMX |
| Alias de gen | ETK/PSCTK2/PSCTK3 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.