Learn More
AADAC, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant AADAC.
Marca: Abnova H00000013-A01.50uL
Descripción
Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens [provided by RefSeq
Sequence: QLLDDPDVKIKLKIQSLIYPALQPLDVDLPSYQENSNFLFLSKSLMVRFWSEYFTTDRSLEKAMLSRQHVPVESSHLFKFINWSSLLPERFIKGHVYNNPEspecificaciones
| AADAC | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant AADAC. | |
| AADAC | |
| CES5A1/DAC | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001086 | |
| AADAC | |
| AADAC (NP_001077, 201 a.a. to 300 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 13 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only