Learn More
Alambre Monel™ 400, 0,50 mm de diámetro, Thermo Scientific Chemicals
Código de producto. p-7059646
Change view
Click to view available options
Cantidad:
113 g
227 g
Tamaño de la unidad:
113 g
227 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
15442127 113 g 113 g
15452127 227 g 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 15442127 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 045066.TB
Solicitar formato a granel o personalizado
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Alloy 400 is widely used in many fields, especially marine and chemical processing. Typical applications are valves and pumps; pump and propeller shafts; marine fixtures and fasteners; electrical and electronic components; springs; chemical processing equipment; gasoline and fresh water tanks; crude petroleum stills, process vessels and piping; boiler feedwater heaters and other heat exchangers; and deaerating heaters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Especificaciones
Nombre del producto químico o material Monel™ 400 wire
Nombre de nota 0.50mm (0.02 in.) dia.
Cantidad 113 g
Peso ≈1.7g/m
Formulario Alambre, ≈1,7g/m
Número MDL MFCD09910536
Información de solubilidad Insoluble in water.