Learn More
Invitrogen™ Monalizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA558730
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Monalizumab is a biosimilar that targets KLRC1.
Especificaciones
| Monalizumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| humZ270; IPH-2201; NN-8765; NNC-0141-0000-0100; NNC-141-01000 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |