Learn More
MON1B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Código de producto. 17561336 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
17561336 20 μL 20 microlitros
17551336 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 17561336 Proveedor Proteintech N.º de proveedor 176381AP20UL
Rabbit Polyclonal Antibody
The MON1B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MON1B. This antibody recognizes human antigen. The MON1B antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Antígeno MON1B
Aplicaciones Western Blot, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 22879
Especies diana Human
Isotype IgG