missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3; MIM 601408), yeast YBF2 and SAS2, and TIP60 (HTATIP; MIM 601409). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.
Especificaciones
Especificaciones
| Antígeno | MOF |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.19 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | KAT8 |
| N.º de referencia del gen | Q5XI06, Q9D1P2, Q9H7Z6 |
| Alias de gen | hMOF, KAT8, MOF, MYST 1, MYST1 |
| Símbolos de los genes | Kat8 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?