Learn More
Moesin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. p-7208295 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16856144 20 μL 20 microlitros
16846144 150 μL 150 microlitros
2 options
Código de producto. 16856144
Marca: Proteintech 164951AP20UL
Rabbit Polyclonal Antibody
This antibody does not react with talin or ezrin. It weakly reacts with radixin. It detects most of the proteolytic fragments of moesin.
Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures. This antibody does not react with talin or ezrin. It weakly reacts with radixin. It detects most of the proteolytic fragments of moesin.
Especificaciones
Antígeno Moesin
Aplicaciones Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Clasificación Polyclonal
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica MSN
N.º de referencia del gen O35763, P26038, P26041
Alias de gen moesin, MSN
Símbolos de los genes Msn
Especie del huésped Rabbit
Inmunógeno Moesin Fusion Protein Ag9623
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17698, 4478, 81521
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG