missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNDA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13576-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with ant
Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with antigenic determits present in normal myeloid cells and leukemias of similar derivation.Especificaciones
| MNDA | |
| Polyclonal | |
| Unconjugated | |
| MNDA | |
| MNDA, PYHIN3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4332 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P41218 | |
| MNDA | |
| MNDA Fusion Protein Ag4476 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido