missing translation for 'onlineSavingsMsg'
Learn More

PeproTech MMP3 Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. 17895483
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17895483 100 μg 100 microgramos
17835793 50 μg 50 microgramos
17855313 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17895483

Marca: PeproTech 500P324100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MRTFPGIPKW RKTHLTYRIV NYTPDLPKDA VDSAVEKALK VWEEVTPLTF SRLYEGEADI MISFAVREHG DFYPFDGPGN VLAHAYAPGP GINGDAHFDD DEQWTKDTTG TNLFLVAAHE IGHSLGLFHS ANTEALMYPL YHSLTDLTRF RLSQDDINGI QSLYGPPPDS PETPLVPTEP VPPEPGTPAN CDPALSFDAV STLRGEILIF KDRHFWRKSL RKLEPELHLI SSFWPSLPSG VDAAYEVTSK DLVFIFKGNQ FWAIRGNEVR AGYPRGIHTL GFPPTVRKID AAISDKEKNK TYFFVEDKYW RFDEKRNSME PGFPKQIAED FPGIDSKIDA VFEEFGFFYF FTGSSQLEFD PNAKKVTHTL KSNSWLNC. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MMP-3. Anti-Human MMP-3-specific antibody was purified by affinity chromatography employing an immobilized Human MMP-3 matrix. Sandwich ELISA: To detect Human MMP-3 by sandwich ELISA (using 100 μL/well), a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MMP-3 (500-P324BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human MMP-3. Western Blot: To detect Human MMP-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human MMP-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MMP3
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative
génica MMP3
N.º de referencia del gen P08254
Alias de gen CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1
Símbolos de los genes MMP3
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human MMP-3.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 4314
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.