Learn More
Novus Biologicals™ MMP-9 Antibody (Yeda patent anti-MMP-9) - Humanized, IgG2SA, Novus Biologicals™
Código de producto. 30047691
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
30047691 100 μg 100 microgramos
30044494 50 μg 50 microgramos
30044485 1 mg 1 miligramo
3 options
Código de producto. 30047691
Marca: Novus Biologicals™ NBP328864100ug
Human Monoclonal Antibody
MMP-9 Monoclonal antibody specifically detects MMP-9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno MMP-9
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon Yeda patent anti-MMP-9
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen P14780
Alias de gen 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Especie del huésped Human
Inmunógeno MMP9
Método de purificación Protein A purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
Primario o secundario Primary
ID de gen (Entrez) 4318
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG2