Learn More
Invitrogen™ Kit de transcripción mMESSAGE mMACHINE™ T3
Obtiene entre 10–50 veces la producción de reacciones de transcripción in vitro convencionales
914.00€
Código de producto | Marca | Tipo de producto | N.º de reacciones | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Tipo de producto | N.º de reacciones | Precio | Cantidad y disponibilidad | ||||
10665605
|
Invitrogen™
AM1348 |
Kit de transcripción in vitro T3 | 25 reacciones |
914.00€
25 reacciones |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Ambion mMESSAGE mMACHINE High Yield Capped RNA Transcription Kits use Ambion's patented high-yield transcription technology for the routine synthesis of 15 - 35μg of 7-methyl guanosine capped RNA from 1μg of template DNA. Each kit includes sufficient reagents for 25 reactions.
- High yields of capped RNA—up to 39μg of RNA per reaction
- Extra convenience—cap analog-nucleotide mix provided
- Simplified reaction format
- Synthesized transcripts ideal for microinjection, in vitro translation, or transfection in a 20μL reaction during a 2 hour incubation, mMESSAGE mMACHINE High Yield Capped RNA Transcription will yield up to 35μg of capped RNA
- ™
- 10–50 times yield obtained with conventional in vitro transcription reactions
- Ratio of cap analog to GTP has been optimized to allow compromise between yield (15 - 35μg) and proportion of transcripts that are capped (80%)
- Contain a LiCl precipitation solution, efficient for separating proteins and unincorporated nucleotides (including free cap analog) from the capped RNA, allowing an increased efficiency of translation
- Optimized for use with polymerases included in kit
- Contains 57U of cap analog