Learn More
MMD2 Polyclonal Antibody
MMD2 Polyclonal Antibody
Marca: Invitrogen PA598604
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MMD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Especificaciones
| MMD2 | |
| Polyclonal | |
| Unconjugated | |
| MMD2 | |
| 4930518M15Rik; C88001; Mmd2; monocyte to macrophage differentiation associated 2; monocyte to macrophage differentiation factor 2; monocyte to macrophage differentiation-associated 2; monocyte-to-macrophage differentiation factor 2; monocyte-to-macrophage differentiation-associated protein 2; PAQR10; Progestin and adipoQ receptor family member 10; progestin and adipoQ receptor family member X | |
| Rabbit | |
| Protein G | |
| RUO | |
| 100505381, 221938, 75104 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IY49, Q8R189 | |
| MMD2 | |
| Recombinant Human Monocyte to macrophage differentiation factor 2 protein (1-38AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |