Learn More
MMAF hydrochloride, MedChemExpress
Código de producto. p-200098883
Encompass_Preferred
Change view
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
Tamaño de la unidad:
10 miligramos
2 miligramos
5 miligramos
16439207 5 mg 5 miligramos
16400256 2 mg 2 miligramos
16426648 10 mg 10 miligramos
3 options
Código de producto. 16439207
Marca: Medchem Express HY15579A
MedChemExpress MMAF (Monomethylauristatin F) hydrochloride is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF hydrochloride is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as Vorsetuzumab mafodotin and SGN-CD19A.
MMAF (Monomethylauristatin F) hydrochloride is a potent tubulin polymerization inhibitor and is used as a antitumor agent. MMAF hydrochloride is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) such as Vorsetuzumab mafodotin and SGN-CD19A.
Especificaciones
Nombre del producto químico o material MMAF hydrochloride
Forma física Solid
CAS 1415246-68-2
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C39H66ClN5O8
Para utilizar con (aplicación) Cancer-programmed cell death
Sinónimo Monomethylauristatin F hydrochloride
Información de solubilidad DMSO : 27 mg/mL (35.14 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC(C)[C@H](NC)C(N[C@H](C(N([C@@H]([C@@H](C)CC)[C@H](OC)CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)=O)C)=O)C(C)C)=O.Cl
Peso molecular (g/mol) 768.42
Formula Weight (peso de la fórmula) 768.42
Porcentaje de pureza 97.71%