missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MLX Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA541683
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: LRKDVTALKI MKVNYEQIVK AHQDNPHEGE DQVSDQVKFN VFQGIMDSLF Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
The product of this gene belongs to the family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors. These factors form heterodimers with Mad proteins and play a role in proliferation, determination and differentiation. This gene product may act to diversify Mad family function by its restricted association with a subset of the Mad family of transcriptional repressors, namely, Mad1 and Mad4. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Especificaciones
| MLX | |
| Polyclonal | |
| Unconjugated | |
| Mlx | |
| bHLHd13; BigMax alpha; BigMax protein; class D basic helix-loop-helix protein 13; hypothetical protein LOC508531; im:7146242; MAD7; MAX dimerization protein MLX; MAX-like bHLHZIP protein; max-like factor protein; max-like protein X; Mlx; MLX, MAX dimerization protein; MXD7; Protein BigMax; TCFL4; TF4; transcription factor like 4; transcription factor-like 4; transcription factor-like protein 4; wu:fb61a01; wu:fb99c01; zgc:112409 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21428, 6945 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O08609, Q9UH92 | |
| Mlx | |
| Synthetic peptide directed towards the C-terminal of human MLX (aa 201-250). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido