Learn More
Invitrogen™ mlaA Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA5144553
Código de producto. 17920223
  • 378.00€ / 100 microlitros
Descripción
mlaA Polyclonal Antibody for Western Blot, ELISA
Involved in a phospholipid transport pathway that maintains lipid asymmetry in the outer membrane by retrograde trafficking of phospholipids from the outer membrane to the inner membrane.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
mlaA
Polyclonal
Unconjugated
mlaA
vacJ
Rabbit
Affinity Chromatography
RUO
945582
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot
4.06 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
P76506
mlaA
Recombinant Escherichia coli (strain K12) mlaA protein (18-251aa).
100 μL
Primary
Bacteria