missing translation for 'onlineSavingsMsg'
Learn More

ML224, MedChemExpress

Código de producto. 16495128
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Product Code. Cantidad unitSize
16495128 10 mg 10 miligramos
16497847 5 mg 5 miligramos
16485846 100 mg 100 miligramos
16441367 50 mg 50 miligramos
4 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 16495128

missing translation for 'mfr': Medchem Express HY12381

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress ML224(NCGC00242364; ANTAG3) is a selective TSHR inverse agonist; inhibits TSH-stimulated cAMP production with an IC50 = 2.3 μM.

ML224(NCGC00242364

Specifications

Nombre del producto químico o material ML224
CAS 1338824-21-7
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C31H31N3O5
Cantidad 10 mg
Sinónimo NCGC00242364 ANTAG3
Información de solubilidad DMSO : 100 mg/mL (190.26 mM; Need ultrasonic)
SMILES CC(NC1=CC(C)=C(OCC2=CC(C(N3CC4=CC=CO4)NC5=C(C=CC=C5)C3=O)=CC=C2OC)C(C)=C1)=O
Peso molecular (g/mol) 525.59
Formula Weight (peso de la fórmula) 525.59
Porcentaje de pureza 99.71%
Grado Research
Forma física Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.